Ethyl Methyl Ketone, Extra Pure, SLR, Fisher Chemical™
Ethyl Methyl Ketone, Extra Pure, SLR, Fisher Chemical™
Ethyl Methyl Ketone, Extra Pure, SLR, Fisher Chemical™
Ethyl Methyl Ketone, Extra Pure, SLR, Fisher Chemical™
Ethyl Methyl Ketone, Extra Pure, SLR, Fisher Chemical™
C4H8O, CAS Number-78-93-3, methyl ethyl ketone, methylethylketone, methyl acetone, 2-butanone, methylethyl ketone, meetco, butanone, 3-butanone, ethyl methyl ketone, ethylmethylketon, 2.5L, 80 deg.C, CHEBI:28398, Colorless, 72.11g/mol, ZWEHNKRNPOVVGH-UHFFFAOYSA-N, butan-2-one, -87 deg.C, 11648
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
2.5 L
5 L
Packaging:
View all
Metal can
Catalog number FKCE145025
also known as FKCE/1450/25
Price (MYR)
2,424.00
EA
Add to cart
Quantity:
25 L
Packaging:
Metal can
Request bulk or custom format
Price (MYR)
2,424.00
EA
Add to cart
Specifications
CAS78-93-3
Melting Point-87°C
ColorColorless
Boiling Point80°C
PackagingMetal can
View more
C4H8O, CAS Number-78-93-3, methyl ethyl ketone, methylethylketone, methyl acetone, 2-butanone, methylethyl ketone, meetco, butanone, 3-butanone, ethyl methyl ketone, ethylmethylketon, 2.5L, 80 deg.C, CHEBI:28398, Colorless, 72.11g/mol, ZWEHNKRNPOVVGH-UHFFFAOYSA-N, butan-2-one, -87 deg.C, 11648