Check Order Status
Ethyl Acetate 99.8+%, for Residue Analysis, Distol™, Fisher Chemical™
Element Search
Structure Search
Ethyl Acetate 99.8+%, for Residue Analysis, Distol™, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
Catalog number
FKCE090317
also known as FKCE/0903/17
Price (MYR)
335.00
BOT
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
335.00
BOT
Add to cart
Specifications
Packaging
Amber glass bottle
CAS
141-78-6
Vapor Pressure
103mbar at 20°C
Viscosity
0.45cp at 20°C
Vapor Density
3.04
View more