Ethyl Acetate, Certified AR for Analysis, Fisher Chemical™
Ethyl Acetate, Certified AR for Analysis, Fisher Chemical™
Ethyl Acetate, Certified AR for Analysis, Fisher Chemical™
Ethyl Acetate, Certified AR for Analysis, Fisher Chemical™
Ethyl Acetate, Certified AR for Analysis, Fisher Chemical™
C4H8O2, CAS Number-141-78-6, acetic acid ethyl ester, acetic acid, ethyl ester, acetic ester, acetic ether, acetidin, acetoxyethane, ethylacetate, ethyl acetic ester, ethyl ethanoate, vinegar naphtha, 2.5L, 75 deg.C, CHEBI:27750, Colorless, 88.11g/mol, XEKOWRVHYACXOJ-UHFFFAOYSA-N, ethyl acetate | CAS: 141-78-6 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
5 L
25 L
Packaging:
View all
Plastic-coated Glass Bottle
Amber Glass Bottle
Catalog number FKCE0900PB17
also known as FKCE/0900/PB17
Price (MYR)
196.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Plastic-coated Glass Bottle
Request bulk or custom format
Price (MYR)
196.00
EA
Add to cart
Specifications
Melting Point-83.5°C
CAS141-78-6
ColorColorless
Boiling Point75°C
Quantity2.5 L
View more
C4H8O2, CAS Number-141-78-6, acetic acid ethyl ester, acetic acid, ethyl ester, acetic ester, acetic ether, acetidin, acetoxyethane, ethylacetate, ethyl acetic ester, ethyl ethanoate, vinegar naphtha, 2.5L, 75 deg.C, CHEBI:27750, Colorless, 88.11g/mol, XEKOWRVHYACXOJ-UHFFFAOYSA-N, ethyl acetate