Check Order Status
Ethyl Acetate, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Ethyl Acetate, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Change view
Quantity:
25 L
Packaging:
Metal Can
Catalog number
FKCE085025
also known as FKCE/0850/25
Price (MYR)
1,242.00
EA
Add to cart
Quantity:
25 L
Packaging:
Metal Can
Request bulk or custom format
Price (MYR)
1,242.00
EA
Add to cart
Specifications
Melting Point
-83.5°C
CAS
141-78-6
Color
Colorless
Boiling Point
75°C
Quantity
25 L
View more
Ethyl Acetate, Extra Pure, SLR