Check Order Status
Ethanolamine, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Ethanolamine, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
Packaging:
Amber glass bottle
Catalog number
FKCE070017
also known as FKCE/0700/17
Price (MYR)
467.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Amber glass bottle
Request bulk or custom format
Price (MYR)
467.00
EA
Add to cart
Specifications
CAS
141-43-5
Melting Point
10°C
Color
Colorless
pH
12
Boiling Point
170°C
View more