Check Order Status
1,4-Dioxane, Certified AR for Analysis, Stabilised with BHT, Fisher Chemical™
Element Search
Structure Search
1,4-Dioxane, Certified AR for Analysis, Stabilised with BHT, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
500 mL
1 L
Packaging:
View all
Plastic-coated Glass Bottle
Catalog number
FKCD4550PB17
also known as FKCD/4550/PB17
Price (MYR)
366.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Plastic-coated Glass Bottle
Request bulk or custom format
Price (MYR)
366.00
EA
Add to cart
Specifications
pH
6 to 8
Melting Point
12°C
CAS
123-91-1
Color
Colorless
Boiling Point
101°C
View more
1,4-Dioxane, Certified AR for Analysis, Stabilised with BHT