Dimethyl Sulfoxide, Extra Pure, SLR, Fisher Chemical™
Dimethyl Sulfoxide, Extra Pure, SLR, Fisher Chemical™
Dimethyl Sulfoxide, Extra Pure, SLR, Fisher Chemical™
Dimethyl Sulfoxide, Extra Pure, SLR, Fisher Chemical™
Dimethyl Sulfoxide, Extra Pure, SLR, Fisher Chemical™
C2H6OS, CAS Number-67-68-5, demsodrox, methyl sulfoxide, dimexide, methane, sulfinylbis, dimethyl sulfoxide, dmso, demavet, dimethylsulfoxide, dimethyl sulphoxide, demasorb, 500mL, 189 deg.C, CHEBI:28262, Colorless, 78.13g/mol, IAZDPXIOMUYVGZ-UHFFFAOYSA-N, methylsulfinylmethane, 18.4 deg.C, 2089
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
HDPE Bottle
Catalog number FKCD4120PB08
also known as FKCD/4120/PB08
Price (MYR)
128.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
HDPE Bottle
Request bulk or custom format
Price (MYR)
128.00
BOT
Add to cart
Specifications
Melting Point18.4°C
CAS67-68-5
ColorColorless
Boiling Point189°C
Quantity500 mL
View more
C2H6OS, CAS Number-67-68-5, demsodrox, methyl sulfoxide, dimexide, methane, sulfinylbis, dimethyl sulfoxide, dmso, demavet, dimethylsulfoxide, dimethyl sulphoxide, demasorb, 500mL, 189 deg.C, CHEBI:28262, Colorless, 78.13g/mol, IAZDPXIOMUYVGZ-UHFFFAOYSA-N, methylsulfinylmethane, 18.4 deg.C, 2089