Check Order Status
DimethylFormamide, for HPLC, Fisher Chemical™
Element Search
Structure Search
DimethylFormamide, for HPLC, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
1 L
Packaging:
Amber glass bottle
Plastic Coated Glass bottle
Catalog number
FKCD384617
also known as FKCD/3846/17
Price (MYR)
212.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Amber glass bottle
Request bulk or custom format
Price (MYR)
212.00
BOT
Add to cart
Specifications
CAS
68-12-2
Melting Point
-61°C
Color
Colorless
Boiling Point
153°C
Packaging
Amber glass bottle
View more
DimethylFormamide, for HPLC