Check Order Status
DimethylFormamide, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
DimethylFormamide, Certified AR for Analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
500 mL
1 L
5 L
Packaging:
View all
Plastic Coated Glass bottle
Amber glass bottle
Catalog number
FKCD3841PB17
also known as FKCD/3841/PB17
Price (MYR)
255.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Plastic Coated Glass bottle
Request bulk or custom format
Price (MYR)
255.00
EA
Add to cart
Specifications
CAS
68-12-2
Melting Point
-61°C
Color
Colorless
Boiling Point
153°C
Packaging
Plastic Coated Glass bottle
View more
DimethylFormamide, Certified AR for Analysis