Check Order Status
DimethylFormamide, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
DimethylFormamide, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
1 L
Packaging:
Amber Glass Bottle
Catalog number
FKCD384017
also known as FKCD/3840/17
Price (MYR)
215.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (MYR)
215.00
BOT
Add to cart
Specifications
Melting Point
-61°C
CAS
68-12-2
Color
Colorless
Boiling Point
153°C
Quantity
2.5 L
View more
DimethylFormamide, Extra Pure, SLR