Check Order Status
Diethyl Ether, for HPLC, Stabilised with Ethanol, Fisher Chemical™
Element Search
Structure Search
Diethyl Ether, for HPLC, Stabilised with Ethanol, Fisher Chemical™
Have Questions?
Change view
Quantity:
1 L
2.5 L
Packaging:
Amber Glass Bottle
Catalog number
FKCD250615
also known as FKCD/2506/15
Price (MYR)
396.00
EA
Add to cart
Quantity:
1 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (MYR)
396.00
EA
Add to cart
Specifications
Melting Point
-116°C
CAS
60-29-7
Color
Colorless
Boiling Point
34.6°C
Quantity
1 L
View more