Check Order Status
Diethyl Ether 99.5+%, for Residue Analysis, Stabilised with Copper Gauze, Distol™, Fisher Chemical™
Element Search
Structure Search
Diethyl Ether 99.5+%, for Residue Analysis, Stabilised with Copper Gauze, Distol™, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
1 L
Catalog number
FKCD250317
also known as FKCD/2503/17
Price (MYR)
598.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
598.00
EA
Add to cart
Specifications
Melting Point
-116°C
CAS
60-29-7
Color
Colorless
Boiling Point
34.6°C
Quantity
2.5 L
View more
Diethyl Ether 99.5+%, for Residue Analysis, Stabilised with Copper Gauze, Distol™