Check Order Status
Diethylamine, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
Diethylamine, Certified AR for Analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
500 mL
Catalog number
FKCD211017
also known as FKCD/2110/17
Price (MYR)
793.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
793.00
EA
Add to cart
Specifications
CAS
109-89-7
Melting Point
-50°C
Color
Colorless
pH
12
Boiling Point
55°C
View more