Check Order Status
Dichloromethane, for HPLC, Stabilised with Amylene, Fisher Chemical™
Element Search
Structure Search
Dichloromethane, for HPLC, Stabilised with Amylene, Fisher Chemical™
CH2Cl2, CAS Number-75-09-2, 2.5L, 39 deg.C, Colorless, 84.93g/mol, -97 deg.C, 881, Amber glass bottle, Liquid, 2.93, 350mbar at 20 deg.C, 0.43 mPaS at 20 deg.C
Have Questions?
Change view
Quantity:
2.5 L
1 L
Packaging:
Amber Glass Bottle
Catalog number
FKCD185617
also known as FKCD/1856/17
Price (MYR)
161.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (MYR)
161.00
BOT
Add to cart
Specifications
Melting Point
-97°C
CAS
75-09-2
Color
Colorless
Boiling Point
39°C
Quantity
2.5 L
View more
CH2Cl2, CAS Number-75-09-2, 2.5L, 39 deg.C, Colorless, 84.93g/mol, -97 deg.C, 881, Amber glass bottle, Liquid, 2.93, 350mbar at 20 deg.C, 0.43 mPaS at 20 deg.C