Dichloromethane 99.8+%, Distol™, Stabilised with Amylene, 2.5L, Fisher Chemical
Dichloromethane 99.8+%, Distol™, Stabilised with Amylene, 2.5L, Fisher Chemical
Dichloromethane 99.8+%, Distol™, Stabilised with Amylene, 2.5L, Fisher Chemical
Dichloromethane 99.8+%, Distol™, Stabilised with Amylene, 2.5L, Fisher Chemical
Dichloromethane 99.8+%, Distol™, Stabilised with Amylene, 2.5L, Fisher Chemical
For residue analysis, Distol™, stabilised with amylene.
Have Questions?
Catalog NumberQuantity
FKCD185317
also known as FKCD/1853/17
2.5 L
Catalog number FKCD185317
also known as FKCD/1853/17
Price (MYR)
268.00
BOT
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
268.00
BOT
Add to cart
Specifications
Melting Point-97°C
CAS75-09-2
ColorColorless
Boiling Point39°C
Quantity2.5 L
View more
  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Impurity profile is also reported using gas chromatography and three different detectors (ECD, FID, and MS) to demonstrate suitable for different analyses
  • Contains approx. 15- 200 ppm amylene as preservative
  • Also meets ACS specifications
  • Intended for research use only