Check Order Status
Dichloromethane 99.8+%, for Residue Analysis, Distol™, Stabilised with Amylene, Fisher Chemical
Element Search
Structure Search
Dichloromethane 99.8+%, for Residue Analysis, Distol™, Stabilised with Amylene, Fisher Chemical
Have Questions?
Catalog number
FKCD185315
also known as FKCD/1853/15
Price (MYR)
177.00
Price (MYR)
177.00
EA
Add to cart
Specifications
Melting Point
-97°C
CAS
75-09-2
Color
Colorless
Boiling Point
39°C
Quantity
1 L
View more