Dichloromethane, Certified AR for Analysis, Stabilised with Amylene, Fisher Chemical™
Dichloromethane, Certified AR for Analysis, Stabilised with Amylene, Fisher Chemical™
Dichloromethane, Certified AR for Analysis, Stabilised with Amylene, Fisher Chemical™
Dichloromethane, Certified AR for Analysis, Stabilised with Amylene, Fisher Chemical™
Dichloromethane, Certified AR for Analysis, Stabilised with Amylene, Fisher Chemical™
CH2Cl2, CAS Number-75-09-2, aerothene mm, methane, dichloro, methane dichloride, methylene bichloride, methylene chloride, methylene dichloride, solaesthin, freon 30, narkotil, solmethine, 2.5L, 39 deg.C, CHEBI:15767, Colorless, 84.93g/mol, YMWUJEATGCHHMB-UHFFFAOYSA-N, dichloromethane, -97 deg.C
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
1 L
2.5 L
5 L
10 L
Packaging:
View all
Metal Can
Catalog number FKCD185225
also known as FKCD/1852/25
Price (MYR)
1,096.00
EA
Add to cart
Quantity:
25 L
Packaging:
Metal Can
Request bulk or custom format
Price (MYR)
1,096.00
EA
Add to cart
Specifications
Melting Point-97°C
CAS75-09-2
ColorColorless
Boiling Point39°C
Quantity25 L
View more
CH2Cl2, CAS Number-75-09-2, aerothene mm, methane, dichloro, methane dichloride, methylene bichloride, methylene chloride, methylene dichloride, solaesthin, freon 30, narkotil, solmethine, 2.5L, 39 deg.C, CHEBI:15767, Colorless, 84.93g/mol, YMWUJEATGCHHMB-UHFFFAOYSA-N, dichloromethane, -97 deg.C