Check Order Status
Cyclohexanone, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
Cyclohexanone, Certified AR for Analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
500 mL
Catalog number
FKCC9050PB17
also known as FKCC/9050/PB17
Price (MYR)
452.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
452.00
EA
Add to cart
Specifications
Melting Point
-47°C
Color
Colorless
Boiling Point
155°C
Quantity
2.5 L
Packaging
Plastic-coated Glass Bottle
View more
Cyclohexanone, Certified AR for Analysis