Check Order Status
Cyclohexane (HPLC), Fisher Chemical™
Element Search
Structure Search
Cyclohexane (HPLC), Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Catalog number
FKCC893617
also known as FKCC/8936/17
Price (MYR)
346.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (MYR)
346.00
BOT
Add to cart
Specifications
Melting Point
6.5°C
CAS
110-82-7
Color
Colorless
Boiling Point
81°C
Quantity
2.5 L
View more