Check Order Status
Cyclohexane 99.5+%, for Residue Analysis, Distol™, Fisher Chemical™
Element Search
Structure Search
Cyclohexane 99.5+%, for Residue Analysis, Distol™, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
1 L
Catalog number
FKCC893317
also known as FKCC/8933/17
Price (MYR)
345.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
345.00
EA
Add to cart
Specifications
Melting Point
6.5°C
CAS
110-82-7
Color
Colorless
Boiling Point
81°C
Quantity
2.5 L
View more