Check Order Status
Cyclohexane, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Cyclohexane, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Change view
Quantity:
500 mL
Packaging:
Plastic Coated Glass bottle
Catalog number
FKCC8920PB08
also known as FKCC/8920/PB08
Price (MYR)
155.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Plastic Coated Glass bottle
Request bulk or custom format
Price (MYR)
155.00
EA
Add to cart
Chemical Identifiers
CAS
110-82-7
Specifications
Packaging
Plastic Coated Glass bottle
CAS
110-82-7
Vapor Pressure
104mbar at 20°C
Viscosity
0.94 mPaS at 20°C
Vapor Density
2.9
View more