Check Order Status
Citric Acid Monohydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., USP, Fisher Chemical™
Element Search
Structure Search
Citric Acid Monohydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., USP, Fisher Chemical™
C6H8O7.H2O, CAS Number-5949-29-1, 5kg, White, 210.15g/mol, 135 deg.C, 149972, Plastic keg, 1.8, Solid
Have Questions?
Change view
Quantity:
1 kg
500 g
2.5 kg
Packaging:
Plastic powder jar
Catalog number
FKCC620060
also known as FKCC/6200/60
Price (MYR)
131.00
EA
Add to cart
Quantity:
1 kg
Packaging:
Plastic powder jar
Request bulk or custom format
Price (MYR)
131.00
EA
Add to cart
Specifications
pH
1.8
CAS
5949-29-1
Melting Point
135°C
Color
White
Molecular Formula
C6H10O8
View more
C6H8O7.H2O, CAS Number-5949-29-1, 5kg, White, 210.15g/mol, 135 deg.C, 149972, Plastic keg, 1.8, Solid