Chloroform, Stabilized with Amylene, for HPLC, Fisher Chemical™
Chloroform, Stabilized with Amylene, for HPLC, Fisher Chemical™
Chloroform, Stabilized with Amylene, for HPLC, Fisher Chemical™
Chloroform, Stabilized with Amylene, for HPLC, Fisher Chemical™
Chloroform, Stabilized with Amylene, for HPLC, Fisher Chemical™
High purity solvent suitable for HPLC, spectrophotometry, and pesticide residue analysis
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
4 L
Packaging:
View all
Amber Glass Bottle
Catalog number FKCC496617
also known as FKCC/4966/17
Price (MYR)
156.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (MYR)
156.00
EA
Add to cart
Specifications
Melting Point-63°C
CAS67-66-3
ColorColorless
Boiling Point61°C
Quantity2.5 L
View more
  • Contains approx. 50ppm amylene as preservative
  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Tested to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
  • Also meets ACS specifications
  • Intended for research use only