Check Order Status
Chloroform, 99.8+%, Certified AR for Analysis, Stabilised with Amylene, Fisher Chemical™
Element Search
Structure Search
Chloroform, 99.8+%, Certified AR for Analysis, Stabilised with Amylene, Fisher Chemical™
CHCl3, CAS Number-67-66-3, 2.5L, 61 deg.C, Colorless, 119.38g/mol, -63 deg.C, 826, Plastic Coated Glass bottle, Liquid, 213mbar at 20 deg.C, 0.56 mPaS at 20 deg.C | CAS: 67-66-3 | CHCl3 | 119.37 g/mol
Have Questions?
Change view
Quantity:
25 L
1 L
2.5 L
Packaging:
View all
Metal Can
Catalog number
FKCC496025
also known as FKCC/4960/25
Price (MYR)
1,193.00
EA
Add to cart
Quantity:
25 L
Packaging:
Metal Can
Request bulk or custom format
Price (MYR)
1,193.00
EA
Add to cart
Specifications
Melting Point
-63°C
CAS
67-66-3
Color
Colorless
Boiling Point
61°C
Quantity
25 L
View more
CHCl3, CAS Number-67-66-3, 2.5L, 61 deg.C, Colorless, 119.38g/mol, -63 deg.C, 826, Plastic Coated Glass bottle, Liquid, 213mbar at 20 deg.C, 0.56 mPaS at 20 deg.C