Check Order Status
Chloroform, 99+%, Extra Pure, Stabilised with Amylene, SLR, Fisher Chemical™
Element Search
Structure Search
Chloroform, 99+%, Extra Pure, Stabilised with Amylene, SLR, Fisher Chemical™
Have Questions?
Change view
Quantity:
25 L
Packaging:
Metal Can
Catalog number
FKCC492025
also known as FKCC/4920/25
Price (MYR)
1,089.00
EA
Add to cart
Quantity:
25 L
Packaging:
Metal Can
Request bulk or custom format
Price (MYR)
1,089.00
EA
Add to cart
Specifications
Melting Point
-63°C
CAS
67-66-3
Color
Colorless
Boiling Point
61°C
Quantity
25 L
View more