Check Order Status
Charcoal, Technical, Decolourising Powder, ActIVated, Fisher Chemical™
Element Search
Structure Search
Charcoal, Technical, Decolourising Powder, ActIVated, Fisher Chemical™
Have Questions?
Change view
Quantity:
500 g
Packaging:
Plastic powder jar
Catalog number
FKCC404053
also known as FKCC/4040/53
Price (MYR)
301.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic powder jar
Request bulk or custom format
Price (MYR)
301.00
EA
Add to cart
Specifications
CAS
7440-44-0
Melting Point
3652°C
Color
Black
Packaging
Plastic powder jar
Molecular Formula
C
View more