Check Order Status
Tert-Butyl Alcohol, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
Tert-Butyl Alcohol, Certified AR for Analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
500 mL
Catalog number
FKCB525117
also known as FKCB/5251/17
Price (MYR)
415.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
415.00
EA
Add to cart
Specifications
pH
7
Melting Point
25°C
CAS
75-65-0
Boiling Point
83°C
Quantity
2.5 L
View more