Check Order Status
Sec-Butyl Alcohol, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
Sec-Butyl Alcohol, Certified AR for Analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
Packaging:
Plastic Coated Glass bottle
Catalog number
FKCB5210PB17
also known as FKCB/5210/PB17
Price (MYR)
-
Quantity:
2.5 L
Packaging:
Plastic Coated Glass bottle
Request bulk or custom format
Specifications
CAS
78-92-2
Melting Point
-115°C
Color
Colorless
Boiling Point
99°C
Packaging
Plastic Coated Glass bottle
View more