Check Order Status
Butan-1-ol, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
Butan-1-ol, Certified AR for Analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
25 L
500 mL
1 L
2.5 L
Packaging:
View all
Metal Can
Catalog number
FKCB485025
also known as FKCB/4850/25
Price (MYR)
1,937.00
EA
Add to cart
Quantity:
25 L
Packaging:
Metal Can
Request bulk or custom format
Price (MYR)
1,937.00
EA
Add to cart
Specifications
Melting Point
-89°C
CAS
71-36-3
Color
Colorless
Boiling Point
117.6°C
Quantity
25 L
View more