Check Order Status
Butan-1-ol, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Butan-1-ol, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Change view
Quantity:
5 L
500 mL
2.5 L
Packaging:
View all
Metal Can
Catalog number
FKCB480021
also known as FKCB/4800/21
Price (MYR)
459.00
EA
Add to cart
Quantity:
5 L
Packaging:
Metal Can
Request bulk or custom format
Price (MYR)
459.00
EA
Add to cart
Specifications
Melting Point
-89°C
CAS
71-36-3
Color
Colorless
Boiling Point
117.6°C
Quantity
5 L
View more
Butan-1-ol, Extra Pure, SLR