Search Thermo Fisher Scientific
Catalog Number | Quantity |
---|---|
FKCA777060 also known as FKCA/7770/60 | 1 kg |
FGCA777050 also known as FGCA/7770/50 | 250 g |
FKCA777050 also known as FKCA/7770/50 | 250 g |
FGCA777060 also known as FGCA/7770/60 | 1 kg |
FGCA777053 also known as FGCA/7770/53 | 500 g |
FKCA777053 also known as FKCA/7770/53 | 500 g |