Check Order Status
Ammonium Acetate, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
Ammonium Acetate, Certified AR for Analysis, Fisher Chemical™
C2H7NO2, CAS Number-631-61-8, 500g, White, 77.08g/mol, 114 deg.C, 13066, Plastic powder jar, Solid
Have Questions?
Change view
Quantity:
5 kg
250 g
500 g
Packaging:
View all
Plastic keg
Catalog number
FKCA344065
also known as FKCA/3440/65
Price (MYR)
783.00
EA
Add to cart
Quantity:
5 kg
Packaging:
Plastic keg
Request bulk or custom format
Price (MYR)
783.00
EA
Add to cart
Specifications
CAS
631-61-8
Melting Point
114°C
Color
White
pH
7
Packaging
Plastic keg
View more
C2H7NO2, CAS Number-631-61-8, 500g, White, 77.08g/mol, 114 deg.C, 13066, Plastic powder jar, Solid