Check Order Status
Ammonia Solution, 25%, Certified AR for Analysis, d=0.91, Fisher Chemical™
Element Search
Structure Search
Ammonia Solution, 25%, Certified AR for Analysis, d=0.91, Fisher Chemical™
H5NO, CAS Number-1336-21-6, 2.5L, 38 deg.C, Colorless, -57 deg.C, 66650, Plastic Coated Glass bottle, 12, Liquid, 0.59, 500 hPa at 20 deg.C
Have Questions?
Change view
Quantity:
2.5 L
1 L
Catalog number
FKCA3360PB17
also known as FKCA/3360/PB17
Price (MYR)
264.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
264.00
EA
Add to cart
Specifications
pH
12
Color
Colorless
Boiling Point
38°C
CAS
1336-21-6
Melting Point
-57°C
View more
H5NO, CAS Number-1336-21-6, 2.5L, 38 deg.C, Colorless, -57 deg.C, 66650, Plastic Coated Glass bottle, 12, Liquid, 0.59, 500 hPa at 20 deg.C