Ammonia Solution, 35%, Certified AR for Analysis, d=0.88, Fisher Chemical™
Ammonia Solution, 35%, Certified AR for Analysis, d=0.88, Fisher Chemical™
Ammonia Solution, 35%, Certified AR for Analysis, d=0.88, Fisher Chemical™
Ammonia Solution, 35%, Certified AR for Analysis, d=0.88, Fisher Chemical™
Ammonia Solution, 35%, Certified AR for Analysis, d=0.88, Fisher Chemical™
H5NO, CAS Number-1336-21-6, aquammonia, ammonium hydroxide solution, ammonium hydroxide nh4 oh, ammonia water, aqua ammonia, ammonia, monohydrate, ammonium hydroxide, ammonia aqueous, household ammonia, ammonia, aqua, 2.5L, CHEBI:18219, Colorless, 35.05g/mol, VHUUQVKOLVNVRT-UHFFFAOYSA-N
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Plastic Coated Glass bottle
Catalog number FKCA3280PB17
also known as FKCA/3280/PB17
Price (MYR)
199.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Plastic Coated Glass bottle
Request bulk or custom format
Price (MYR)
199.00
EA
Add to cart
Specifications
pH12
ColorColorless
CAS1336-21-6
Molecular FormulaH5NO
MDL NumberMFCD00066650
View more
H5NO, CAS Number-1336-21-6, aquammonia, ammonium hydroxide solution, ammonium hydroxide nh4 oh, ammonia water, aqua ammonia, ammonia, monohydrate, ammonium hydroxide, ammonia aqueous, household ammonia, ammonia, aqua, 2.5L, CHEBI:18219, Colorless, 35.05g/mol, VHUUQVKOLVNVRT-UHFFFAOYSA-N