Check Order Status
Acetonitrile, for HPLC-MS
Element Search
Structure Search
Acetonitrile, for HPLC-MS
Have Questions?
Change view
Quantity:
2.5 L
1 L
Catalog number
FKCA063817
also known as FKCA/0638/17
Price (MYR)
594.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
594.00
EA
Add to cart
Specifications
Packaging
Amber glass bottle
CAS
75-05-8
Vapor Pressure
97mbar at 20°C
Viscosity
0.36cp at 20°C
Vapor Density
1.42
View more
Acetonitrile, for HPLC-MS