Check Order Status
Acetonitrile 99.8+%, for Residue Analysis, Distol™, Fisher Chemical™
Element Search
Structure Search
Acetonitrile 99.8+%, for Residue Analysis, Distol™, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
Catalog number
FKCA062317
also known as FKCA/0623/17
Price (MYR)
564.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
564.00
EA
Add to cart
Specifications
Melting Point
-46°C
CAS
75-05-8
Color
Colorless
Boiling Point
81°C
Quantity
2.5 L
View more