Check Order Status
Acetonitrile, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
Acetonitrile, Certified AR for Analysis, Fisher Chemical™
C2H3N, CAS Number-75-05-8, 5L, 81 deg.C, Colorless, 41.05g/mol, -46 deg.C, 1878, Metal can, Liquid, 1.42, 97mbar at 20 deg.C, 0.36cp at 20 deg.C
Have Questions?
Change view
Quantity:
2.5 L
1 L
Packaging:
Plastic-coated Glass Bottle
Catalog number
FKCA0621PB17
also known as FKCA/0621/PB17
Price (MYR)
353.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Plastic-coated Glass Bottle
Request bulk or custom format
Price (MYR)
353.00
BOT
Add to cart
Specifications
Melting Point
-46°C
CAS
75-05-8
Color
Colorless
Boiling Point
81°C
Quantity
2.5 L
View more
C2H3N, CAS Number-75-05-8, 5L, 81 deg.C, Colorless, 41.05g/mol, -46 deg.C, 1878, Metal can, Liquid, 1.42, 97mbar at 20 deg.C, 0.36cp at 20 deg.C