Check Order Status
Acetone, for Trace Metal Analysis, Primar™, Fisher Chemical
Element Search
Structure Search
Acetone, for Trace Metal Analysis, Primar™, Fisher Chemical
Have Questions?
Catalog Number
Quantity
FKCA060417
also known as FKCA/0604/17
2.5 L
Catalog number
FKCA060417
also known as FKCA/0604/17
Price (MYR)
362.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
362.00
EA
Add to cart
Chemical Identifiers
CAS
67-64-1
Specifications
Packaging
Amber glass bottle
CAS
67-64-1
Vapor Pressure
247mbar at 20°C
Viscosity
0.32 mPaS at 20°C
Vapor Density
2
View more