Check Order Status
Acetone 99.8+%, for Residue Analysis, Distol™, Fisher Chemical™
Element Search
Structure Search
Acetone 99.8+%, for Residue Analysis, Distol™, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
1 L
Catalog number
FKCA060317
also known as FKCA/0603/17
Price (MYR)
303.00
BOT
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
303.00
BOT
Add to cart
Specifications
Packaging
Amber glass bottle
CAS
67-64-1
Vapor Pressure
247mbar at 20°C
Viscosity
0.32 mPaS at 20°C
Vapor Density
2
View more