Red SIRCHMARK™ Evidence Tape
Red SIRCHMARK™ Evidence Tape
Actual product may vary
Red SIRCHMARK™ Evidence Tape
Simply peel the evidence strip away from the liner and apply to evidence.
Have Questions?
Catalog number FISS99539
Price (MYR)
-
  • Translucent red with opaque black overprinting
  • Printable tape allows you to initial and date items as necessary
  • 54-foot roll
  • Coated with a permanent, quick-setting acrylic adhesive that defies removal from most surfaces
  • Roll is 1.5 in. wide, 0.001 in thick, on a 3 in. core and is backed with 50 lb. stock Kraft liner
  • Attempts to remove SIRCHMARK evidence tape will cause the tape substrate to break and peel off in irregular strips
Specifications
Product TypeTape
Dimensions (L x W)54 ft. x 1.5 in. (16.4m x 3.7cm)
For Use With (Application)Hospitals and laboratories, banks and race tracks applications
MaterialAcetate
RequiresTamperproof seal
Unit SizeEA