Horizontal Pipette Stand
Horizontal Pipette Stand
Actual product may vary
Horizontal Pipette Stand
This Horizontal Pipette Stand is polypropylene and autoclavable.
Have Questions?
Catalog NumberPositions
FISS9937512
Catalog number FISS99375
Price (MYR)
-
Positions:
12
  • Can hold 12 pipettes, 6 on each side, in a horizontal position
  • Stand has two sides which are attach to each other with three metallic rods coated with polypropylene
Specifications
Product TypePipet Rack
Array2 x 6
AutoclavableAutoclavable
ColorTranslucent
HoldsPipets
MaterialPolypropylene
Positions12
ShapeTriangle
ShelvesAngled
StackableYes, When Empty
SterilityNon-sterile
Width (Metric)15 cm
Height (Metric)15 cm
Unit SizeEA