Hall's Car Pair
Hall's Car Pair
Actual product may vary
Hall's Car Pair
Essential items for dynamics experiments.
Have Questions?
Catalog number FISS96317
Price (MYR)
-
Use in conjunction with inclined plane, pulley and weights to study relationship between work and energy.
  • Low-friction wheels
  • Oil-free sleeve bearings — never needs adjustment
  • Durable plastic molded body has deep well for weights and low profile
Specifications
Product TypeHall'S Car
For Use With (Application)For use in dynamics experiments and use to study relationship between work and energy
For Use With (Equipment)Use in conjunction with inclined plane, pulley and weights
Unit SizePAA