Basic Inclined Plane
Basic Inclined Plane
Actual product may vary
Basic Inclined Plane
Designed specially for students.
Have Questions?
Catalog number FISS96183
Price (MYR)
-
For small-scale demonstrations.
  • Measures vertical displacement, angular displacement and distance with built-in scales
  • Heavy-gauge steel construction
Specifications
Product TypeInclined Plane
MaterialHeavy Gauge Steel
For Use With (Application)For small scale demonstrations measures vertical displacement, angular displacement and distance with built in scales
IncludesInstructions and accessories
Dimensions (L x W x H)24.4 x 7.48 x 2.36 in. (62 x 19 x 6 cm)
Height (English)2.36 in.
Height (Metric)6 cm
Length (English)24.4 in.
Length (Metric)62 cm
Width (English)7.48 in.
Width (Metric)19 cm
Weight (English)6.37 lb.
Weight (Metric)2890 g
Unit SizeEA