▸
Funnels
▸
Filtering Funnels
▸
Glass Funnel Filters
Actual product may vary
Glass Funnel Filters
Eisco™ Glass Funnel Filters are made from high quality borosilicate glass and designed with an angled stem and beaded rim for durability.
Have Questions?
Catalog Number
Diameter (Metric)Top
Diameter (Metric) Stem
Length (Metric) Stem
FISS89478
125 mm
16 mm
11.43 cm
Catalog number
FISS89478
Price (MYR)
0.00
Each
Add to cart
Diameter (Metric)Top:
125 mm
Diameter (Metric) Stem:
16 mm
Length (Metric) Stem:
11.43 cm
Price (MYR)
0.00
Each
Add to cart
Funnel filters made from high quality borosilicate glass
60° angle at bottom of stem
Autoclavable
Specifications
Diameter (Metric)Top
125 mm
Capacity (Metric)
1000 mL
Diameter (Metric) Stem
16 mm
Material
Borosilicate Glass
Height (Metric)
22 cm
Angle
60°
Autoclavable
Autoclavable
Color
Clear
Graduated
No
For Use With (Application)
Transferring liquids
Length (Metric) Stem
11.43 cm
Quantity
1 Ea.
Height (English)
8.7 in.
Product Type
Filter Funnel