Kipp's Gas Generator
Kipp's Gas Generator
Actual product may vary
Fisherbrand
Kipp's Gas Generator
Kipp's Gas Generator used to produce controllable amounts of common gases, such as hydrogen and carbon dioxide.
Have Questions?
Catalog NumberCapacity
FISS89312250 mL
Catalog number FISS89312
Price (MYR)
-
Capacity:
250 mL
  • Kipp's gas generator with ground glass connections.
  • Made from high quality borosilicate 3.3 glass.
  • Used to produce small amounts of common gases such as hydrogen, carbon dioxide, etc.
  • Autoclavable.
  • Specifications
    Height (English)5.1 in.
    Height (Metric)13 cm
    Width (English)1 in.
    Width (Metric)2.5 cm
    Capacity250 mL
    For Use With (Application)Create gas
    TypeGas Generator
    Unit SizeEA