Glass Eudiometer Tube with Platinum Electrodes
Glass Eudiometer Tube with Platinum Electrodes
Actual product may vary
Fisherbrand
Glass Eudiometer Tube with Platinum Electrodes
Eudiometer Tubes are closed at one end and include two sealed platinum electrodes. Ideal for measuring the volume gas produced or consumed in a chemical reaction.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FISS89304100 mL
FISS2734650 mL
Catalog number FISS89304
Price (MYR)
-
Capacity (Metric):
100 mL
  • For measuring the volume of gas produced or consumed in a chemical reaction.
  • Constructed of high quality borosilicate glass.
  • Two platinum electrodes are sealed.
  • Tube is closed at one end.
  • White, 0.20 mL graduations.
Specifications
Product TypeEudiometer Tube
ColorClear
DescriptionPlatinum electrodes
Grade7 to Undergraduate
Age12 to 20
For Use With (Application)Measuring the volume of gas produced or consumed in a chemical reaction
Includes1 Tube encasing 2 Platinum Elecrodes
MaterialBorosilicate Glass
Quantity1 Ea.
Weight (English)0.5 lb.
Weight (Metric)226.79 g
Length (English)27 in.
Length (Metric)685.8 mm
Capacity (Metric)100 mL
Class Size1 Lab Group
Unit SizeEA