Solvent Spill Kit
Actual product may vary
Fisherbrand
Solvent Spill Kit
Contain, remove and tag spills fast.
Have Questions?
Catalog NumberCapacity (English)
FISS80201C0.26 gal.
Catalog number FISS80201C
Price (MYR)
-
Capacity (English):
0.26 gal.
  • One instruction sheet/SDS
  • Two gloves
  • Two 9 x 16 in. (22.9 x 40.6 cm) polybags
  • Two twist ties
  • Two blank shipping tags
  • One dust pan and brush
  • One bag diatomaceous earth
Specifications
ColorWhite
Capacity (English)0.26 gal.
Capacity (Metric)1 L
MaterialPolyethylene, Polypropylene
TypeChemical Spill Sorbent Response Pack
For Use With (Application)For labs
Includes1 instruction sheet/MSDS, 2 vinyl exam gloves, two 9 x 16 in. polybags, 2 twist ties, 2 blank shipping tags, 1 dust pan and brush, 1 bag diatomaceous earth
Packaging Quantity1 Ea.
Dimensions (L x W x H)10 x 8 x 8 in.
Weight (English)2.37 lb.
Weight (Metric)1.08 kg
Unit SizeEach