Acid Spill Cleanup Kit
Acid Spill Cleanup Kit
Actual product may vary
Fisherbrand
Acid Spill Cleanup Kit
Contain, remove and tag spills fast.
Have Questions?
Catalog NumberCapacity (English)
FISS80201A0.26 gal.
Catalog number FISS80201A
Price (MYR)
-
Capacity (English):
0.26 gal.
  • One instruction sheet/SDS
  • Two gloves
  • Two 9 x 16 in. (22.9 x 40.6 cm) polybags
  • Two twist ties
  • Two blank shipping tags
  • One dust pan and brush
  • One bag vermiculite absorbent
  • One bottle neutralizing mixture (Calcium Hydroxide, Sodium Carbonate, Calcium Carbonate and Litmus Powder)
WARNING: This product can expose you to chemicals including Respirable crystalline silica, which is known to the State of California to cause cancer. For more information go to http://www.P65Warnings.ca.gov.
Specifications
ColorWhite
Capacity (English)0.26 gal.
Capacity (Metric)1 L
MaterialPolyethylene, Polypropylene
TypeAcid Neutralizing Spill Kit
For Use With (Application)For labs
Includes1 instruction sheet/MSDS, 2 vinyl exam gloves, two 9 x 16 in. polybags, 2 twist ties, 2 blank shipping tags, 1 dust pan and brush, 1 bag vermiculite absorbent, 1 bag neutralizing mixture
Packaging Quantity1 Ea.
Weight (English)4.28 lb.
Weight (Metric)1.94 kg
Unit SizeEA