Featherweight Entomology Forceps, Set of 2
Featherweight Entomology Forceps, Set of 2
Actual product may vary
Featherweight Entomology Forceps, Set of 2
This Entomology Forceps Set includes one narrow-point forceps and one broad-point forceps, specifically designed for precise work in the field of entomology.
Have Questions?
Catalog number FISS72110
Price (MYR)
-
  • These Featherweight Entomology Forceps, available in a set of two, offer exceptional flexibility and can be bent into various shapes to suit your needs.
  • They are especially well-suited for handling delicate specimens such as larvae, eggs, or those stored in vials, ensuring that your specimens remain intact without risk of crushing.
Specifications
Product TypeEntomology Forceps
MaterialStainless Steel
IncludesSet of 2 forceps - one fine tip, one wide tip
Length (English)Wide tip: 4.25 in.; Fine tip: 4.5 in.
Unit SizeEA