Specifications
Product TypeTube Rack
DescriptionFisherbrand HDPE Coated Wire Rack
Array4 x 12
AutoclavableAutoclavable
ColorBlue
HoldsTubes
MaterialHDPE-coated Wire
Positions48
ShapeRectangular
ShelvesNone
StackableNo
SterilityNon-sterile
For Use With (Application)Holds Tubes
No. per Case56/Cs.
No. per Pack1 Ea.
Temperature (Metric) Range-40°C and 121°C
Dimensions (L x W x H)9.40 x 3.30 x 2.40 in.
Length (Metric)23.88 cm
Length (English)9.40 in.
Width (Metric)8.38 cm
Width (English)3.30 in.
Height (Metric)6.10 cm
Height (English)2.40 in.
Diameter (Metric) Well13 to 16 mm
Weight (English)0.3 lb.
Weight (Metric)0.14 kg
Unit SizeEA